accessibility menu, dialog, popup

UserName

CH3H3CCH3N

CAS RN 4210-32-6

IUPAC Name: 4-tert-butylbenzonitrile
Synonyms: 4-tert-butylbenzonitrile
Molecular Weight (g/mol): 159.23
Molecular Formula: C11H13N
InChi Key: IIZURLNRIMKEDL-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C#N
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)