Learn More
4-tert-Butylbenzonitrile, 98+%
CAS: 4210-32-6 | C11H13N | 159.232 g/mol
$69.56 - $550.78
Chemical Identifiers
| CAS | 4210-32-6 |
|---|---|
| Molecular Formula | C11H13N |
| Molecular Weight (g/mol) | 159.232 |
| MDL Number | MFCD00075840 |
| InChI Key | IIZURLNRIMKEDL-UHFFFAOYSA-N |
| Synonym | 4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d |
| PubChem CID | 77883 |
| IUPAC Name | 4-tert-butylbenzonitrile |
| SMILES | CC(C)(C)C1=CC=C(C=C1)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1407603
|
Thermo Scientific Chemicals
A1407603 |
1 g |
Each for $69.56
|
|
|||||
|
AAA1407606
|
Thermo Scientific Chemicals
A1407606 |
5 g |
Each for $151.64
|
|
|||||
|
AAA1407614
|
Thermo Scientific Chemicals
A1407614 |
25 g |
Each for $550.78
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4210-32-6 | |
| 159.232 | |
| IIZURLNRIMKEDL-UHFFFAOYSA-N | |
| 77883 | |
| CC(C)(C)C1=CC=C(C=C1)C#N |
| C11H13N | |
| MFCD00075840 | |
| 4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d | |
| 4-tert-butylbenzonitrile |
Specifications
| 4210-32-6 | |
| 258°C | |
| Characteristic | |
| 1.517 | |
| 1 g | |
| 4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d | |
| CC(C)(C)C1=CC=C(C=C1)C#N | |
| 159.232 | |
| 159.23 | |
| 4-tert-Butylbenzonitrile |
| 0.94 | |
| >110°C (230°F) | |
| C11H13N | |
| MFCD00075840 | |
| 2080049 | |
| IIZURLNRIMKEDL-UHFFFAOYSA-N | |
| 4-tert-butylbenzonitrile | |
| 77883 | |
| ≥98% |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 224-137-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only