missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzonitrile, 98+%

Catalog No. AAA1407603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1407603 1 g
AAA1407606 5 g
AAA1407614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1407603 Supplier Thermo Scientific Chemicals Supplier No. A1407603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4210-32-6 | C11H13N | 159.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4210-32-6
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.232
MDL Number MFCD00075840
InChI Key IIZURLNRIMKEDL-UHFFFAOYSA-N
Synonym 4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d
PubChem CID 77883
IUPAC Name 4-tert-butylbenzonitrile
SMILES CC(C)(C)C1=CC=C(C=C1)C#N

Specifications

Density 0.94
Boiling Point 258°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.517
Quantity 1 g
Beilstein 2080049
Formula Weight 159.23
Percent Purity ≥98%
Chemical Name or Material 4-tert-Butylbenzonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 224-137-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only