missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzenethiol 97.0+%, TCI America™

Catalog No. p-7118651
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B07415G 5 g
B074125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. B07415G missing translation for 'mfr' TCI America missing translation for 'supplierNo' B07415G
Only null left

Chemical Identifiers

CAS 2396-68-1
Molecular Formula C10H14S
Molecular Weight (g/mol) 166.282
MDL Number MFCD00022067
InChI Key GNXBFFHXJDZGEK-UHFFFAOYSA-N
Synonym 4-tert-butylthiophenol, p-tert-butylthiophenol, benzenethiol, p-tert-butyl, p-tert-butylbenzenethiol, p-t-butylthiophenol, p-tert-butylphenyl mercaptan, benzenethiol, 4-1,1-dimethylethyl, benzenethiol, p-tert-butylthio, 4-tert-butyl thiophenol, 4-t-butylthiophenol
PubChem CID 75454
IUPAC Name 4-tert-butylbenzenethiol
SMILES CC(C)(C)C1=CC=C(C=C1)S

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 166.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzenethiol
EINECSNumber (3)-1093
RTECSNumber DC1025000
TSCA No