missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzaldehyde 95.0+%, TCI America™

Catalog No. B176025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B176025G 25 g
B1760100G 100 g
B1760500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B176025G Supplier TCI America Supplier No. B176025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 939-97-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00035742
InChI Key OTXINXDGSUFPNU-UHFFFAOYSA-N
Synonym 4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl
PubChem CID 70324
IUPAC Name 4-tert-butylbenzaldehyde
SMILES CC(C)(C)C1=CC=C(C=O)C=C1

Specifications

Color Yellow
Boiling Point 246°C
Quantity 25 g
UN Number 3082
Formula Weight 162.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzaldehyde
EINECSNumber (3)-2663
TSCA Yes