Learn More
4-tert-Butylbenzaldehyde, 95%
CAS: 939-97-9 | C11H14O | 162.23 g/mol
$154.94 - $154.94
Chemical Identifiers
| CAS | 939-97-9 |
|---|---|
| Molecular Formula | C11H14O |
| Molecular Weight (g/mol) | 162.23 |
| MDL Number | MFCD00035742 |
| InChI Key | OTXINXDGSUFPNU-UHFFFAOYSA-N |
| Synonym | 4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl |
| PubChem CID | 70324 |
| IUPAC Name | 4-tert-butylbenzaldehyde |
| SMILES | CC(C)(C)C1=CC=C(C=O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC426520500
|
Thermo Scientific Chemicals
426520500 |
50 mL | Glass bottle |
Each for $154.94
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 939-97-9 | |
| 162.23 | |
| OTXINXDGSUFPNU-UHFFFAOYSA-N | |
| 70324 | |
| CC(C)(C)C1=CC=C(C=O)C=C1 |
| C11H14O | |
| MFCD00035742 | |
| 4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl | |
| 4-tert-butylbenzaldehyde |
Specifications
| 939-97-9 | |
| 100.0 | |
| 0.9700g/mL | |
| 101°C | |
| 94% min. (GC) | |
| C11H14O | |
| (CH3)3CC6H4CHO | |
| 50 mL | |
| 4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl | |
| OTXINXDGSUFPNU-UHFFFAOYSA-N | |
| 4-tert-butylbenzaldehyde | |
| 70324 | |
| 162.23 | |
| Liquid |
| 94.0 | |
| Colorless to Yellow | |
| 249°C | |
| Authentic | |
| Glass bottle | |
| 1.527 | |
| MFCD00035742 | |
| 0.97 | |
| Solubility in water: insoluble | |
| CC(C)(C)C1=CC=C(C=O)C=C1 | |
| 162.23 | |
| 0.004 Pas (20°C) | |
| 95% | |
| 4-tert-Butylbenzaldehyde |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 213-367-9
RUO – Research Use Only