missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzaldehyde, 95%

Catalog No. AC426520500
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426520500 50 mL Glass bottle
AC426522500 250 mL Glass bottle
AC426520010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC426520500 Supplier Thermo Scientific Chemicals Supplier No. 426520500

CAS: 939-97-9 | C11H14O | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 939-97-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00035742
InChI Key OTXINXDGSUFPNU-UHFFFAOYSA-N
Synonym 4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl
PubChem CID 70324
IUPAC Name 4-tert-butylbenzaldehyde
SMILES CC(C)(C)C1=CC=C(C=O)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 249°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.527
Linear Formula (CH3)3CC6H4CHO
Quantity 50 mL
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble
Viscosity 0.004 Pas (20°C)
Formula Weight 162.23
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-367-9

RUO – Research Use Only