missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butyl-o-xylene 98.0+%, TCI America™
Supplier: TCI America B133825G
Specifications
| 4-tert-Butyl-o-xylene | |
| Colorless | |
| C12H18 | |
| 25 g | |
| QRPPSTNABSMSCS-UHFFFAOYSA-N | |
| 4-tert-butyl-1,2-dimethylbenzene | |
| 81881 | |
| ≥98.0% (GC) |
| 7397-06-0 | |
| 211°C | |
| MFCD00038277 | |
| 4-tert-butyl-o-xylene, 4-tert-butyl-1,2-dimethylbenzene, 4-t-butyl-o-xylene, benzene, 4-1,1-dimethylethyl-1,2-dimethyl, acmc-209ot4, o-xylene, 4-tert-butyl, as-p2, 1,2-dimethyl-4-t-butylbenzene, 3,4-dimethyl-tert-butylbenzene | |
| CC1=C(C=C(C=C1)C(C)(C)C)C | |
| 162.276 | |
| 162.28 | |
| Liquid |
Chemical Identifiers
| 7397-06-0 | |
| 162.276 | |
| QRPPSTNABSMSCS-UHFFFAOYSA-N | |
| 81881 | |
| CC1=C(C=C(C=C1)C(C)(C)C)C |
| C12H18 | |
| MFCD00038277 | |
| 4-tert-butyl-o-xylene, 4-tert-butyl-1,2-dimethylbenzene, 4-t-butyl-o-xylene, benzene, 4-1,1-dimethylethyl-1,2-dimethyl, acmc-209ot4, o-xylene, 4-tert-butyl, as-p2, 1,2-dimethyl-4-t-butylbenzene, 3,4-dimethyl-tert-butylbenzene | |
| 4-tert-butyl-1,2-dimethylbenzene |
Safety and Handling
TSCA : Yes