CAS RN 7397-06-0
IUPAC Name:
4-tert-butyl-1,2-dimethylbenzene
Synonyms:
4-tert-butyl-1,2-dimethylbenzene
Molecular Weight (g/mol):
162.28
Molecular Formula:
C12H18
InChi Key:
QRPPSTNABSMSCS-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1C)C(C)(C)C
1
–
1
of
1
results
1
4-tert-Butyl-o-xylene 98.0+%, TCI America™
CAS: 7397-06-0 Molecular Formula: C12H18 Molecular Weight (g/mol): 162.276 MDL Number: MFCD00038277 InChI Key: QRPPSTNABSMSCS-UHFFFAOYSA-N Synonym: 4-tert-butyl-o-xylene,4-tert-butyl-1,2-dimethylbenzene,4-t-butyl-o-xylene,benzene, 4-1,1-dimethylethyl-1,2-dimethyl,acmc-209ot4,o-xylene, 4-tert-butyl,as-p2,1,2-dimethyl-4-t-butylbenzene,3,4-dimethyl-tert-butylbenzene PubChem CID: 81881 IUPAC Name: 4-tert-butyl-1,2-dimethylbenzene SMILES: CC1=C(C=C(C=C1)C(C)(C)C)C