missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)benzylchloride, 97%

Catalog No. p-78833
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC345490050 5 g Glass bottle
AC345490250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC345490050 Supplier Thermo Scientific Chemicals Supplier No. 345490050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19692-45-6 | C11H15Cl | 182.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19692-45-6
Molecular Formula C11H15Cl
Molecular Weight (g/mol) 182.69
MDL Number MFCD00000918
InChI Key WAXIFMGAKWIFDQ-UHFFFAOYSA-N
Synonym 4-tert-butylbenzyl chloride, 1-tert-butyl-4-chloromethyl benzene, p-tert-butylbenzyl chloride, 4-tert-butyl benzyl chloride, benzene, 1-chloromethyl-4-1,1-dimethylethyl, 4-tert-butyl benzylchloride, p-t-butylbenzyl chloride, 1-chloromethyl-4-tert-butylbenzene, p-tert-butyl-.alpha.-chlorotoluene
PubChem CID 88198
IUPAC Name 1-tert-butyl-4-(chloromethyl)benzene
SMILES CC(C)(C)C1=CC=C(CCl)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 98°C to 103°C (7.0 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5205 to 1.5225
Linear Formula (CH3)3CC6H4CH2Cl
Quantity 5 g
Specific Gravity 0.94
Formula Weight 182.69
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(tert-Butyl)benzylchloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 243-228-8
TSCA TSCA

RUO – Research Use Only