missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl-3-methoxy-2,6-dinitrotoluene 99.0+%, TCI America™

$108.11 - $579.71

Chemical Identifiers

CAS 83-66-9
Molecular Formula C12H16N2O5
Molecular Weight (g/mol) 268.269
MDL Number MFCD00024268
InChI Key SUAUILGSCPYJCS-UHFFFAOYSA-N
Synonym 6-tert-Butyl-3-methyl-2,4-dinitroanisole
PubChem CID 6753
ChEBI CHEBI:82495
IUPAC Name 1-tert-butyl-2-methoxy-4-methyl-3,5-dinitrobenzene
SMILES CC1=C(C=C(C(=C1[N+](=O)[O-])OC)C(C)(C)C)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B137925G
View Documents
TCI America
B137925G
25 g
Each for $108.11
Only null left
 
B1379500G
View Documents
TCI America
B1379500G
500 g
Each for $579.71
Only null left
 
Description

Description

Specifications

Chemical Identifiers

83-66-9
268.269
SUAUILGSCPYJCS-UHFFFAOYSA-N
6753
1-tert-butyl-2-methoxy-4-methyl-3,5-dinitrobenzene
C12H16N2O5
MFCD00024268
6-tert-Butyl-3-methyl-2,4-dinitroanisole
CHEBI:82495
CC1=C(C=C(C(=C1[N+](=O)[O-])OC)C(C)(C)C)[N+](=O)[O-]

Specifications

83-66-9
Yellow
C12H16N2O5
25 g
SUAUILGSCPYJCS-UHFFFAOYSA-N
1-tert-butyl-2-methoxy-4-methyl-3,5-dinitrobenzene
6753
268.27
Crystalline Powder
86°C
185°C
MFCD00024268
6-tert-Butyl-3-methyl-2,4-dinitroanisole
CC1=C(C=C(C(=C1[N+](=O)[O-])OC)C(C)(C)C)[N+](=O)[O-]
268.269
CHEBI:82495
≥99.0% (GC)
4-tert-Butyl-3-methoxy-2,6-dinitrotoluene
Videos
Safety and Handling

Safety and Handling

RTECSNumber : BZ8575000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'