missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl-3-methoxy-2,6-dinitrotoluene 99.0+%, TCI America™

Supplier:  TCI America B137925G

 View more versions of this product

Catalog No. B137925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-tert-Butyl-3-methoxy-2,6-dinitrotoluene
86°C
185°C
MFCD00024268
6-tert-Butyl-3-methyl-2,4-dinitroanisole
CC1=C(C=C(C(=C1[N+](=O)[O-])OC)C(C)(C)C)[N+](=O)[O-]
268.269
CHEBI:82495
≥99.0% (GC)
83-66-9
Yellow
C12H16N2O5
25 g
SUAUILGSCPYJCS-UHFFFAOYSA-N
1-tert-butyl-2-methoxy-4-methyl-3,5-dinitrobenzene
6753
268.27
Crystalline Powder

Chemical Identifiers

83-66-9
268.269
SUAUILGSCPYJCS-UHFFFAOYSA-N
6753
1-tert-butyl-2-methoxy-4-methyl-3,5-dinitrobenzene
C12H16N2O5
MFCD00024268
6-tert-Butyl-3-methyl-2,4-dinitroanisole
CHEBI:82495
CC1=C(C=C(C(=C1[N+](=O)[O-])OC)C(C)(C)C)[N+](=O)[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : BZ8575000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions