missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl-2-nitrophenol 98.0+%, TCI America™

Catalog No. p-7119281
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18125G 5 g
B181225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18125G Supplier TCI America Supplier No. B18125G

Chemical Identifiers

CAS 3279-07-0
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00191888
InChI Key IHGNADPMUSNTJW-UHFFFAOYSA-N
Synonym 2-nitro-4-tert-butylphenol, 4-tert-butyl-2-nitrophenol, phenol, 4-1,1-dimethylethyl-2-nitro, 4-tert.-butyl-2-nitrophenol, acmc-209hvt, o-nitro-p-t-butyl-phenol, 2-nitro-4-t-butyl phenol, 4-t-butyl-2-nitro-phenol, 4-t-butyl-2-nitrophenol
PubChem CID 18661
IUPAC Name 4-tert-butyl-2-nitrophenol
SMILES CC(C)(C)C1=CC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Quantity 5 g
Formula Weight 195.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butyl-2-nitrophenol
EINECSNumber (3)-4600
TSCA Yes