CAS RN 3279-07-0
IUPAC Name:
4-tert-butyl-2-nitrophenol
Synonyms:
4-(tert-butyl)-2-nitrophenol
Molecular Weight (g/mol):
195.22
Molecular Formula:
C10H13NO3
InChi Key:
IHGNADPMUSNTJW-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(O)C(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
4-tert-Butyl-2-nitrophenol 98.0+%, TCI America™
CAS: 3279-07-0 Molecular Formula: C10H13NO3 Molecular Weight (g/mol): 195.218 MDL Number: MFCD00191888 InChI Key: IHGNADPMUSNTJW-UHFFFAOYSA-N Synonym: 2-nitro-4-tert-butylphenol,4-tert-butyl-2-nitrophenol,phenol, 4-1,1-dimethylethyl-2-nitro,4-tert.-butyl-2-nitrophenol,acmc-209hvt,o-nitro-p-t-butyl-phenol,2-nitro-4-t-butyl phenol,4-t-butyl-2-nitro-phenol,4-t-butyl-2-nitrophenol PubChem CID: 18661 IUPAC Name: 4-tert-butyl-2-nitrophenol SMILES: CC(C)(C)C1=CC(=C(C=C1)O)[N+](=O)[O-]