missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butyl-2,6-dimethylphenylsulfur Trifluoride 90.0+%, TCI America™
$96.26 - $279.15
Chemical Identifiers
| CAS | 947725-04-4 |
|---|---|
| Molecular Formula | C12H17F3S |
| Molecular Weight (g/mol) | 250.323 |
| MDL Number | MFCD11858620 |
| InChI Key | VRTQPEYVMHATOA-UHFFFAOYSA-N |
| Synonym | FLUOLEAD(TM) |
| PubChem CID | 44184700 |
| IUPAC Name | (4-tert-butyl-2,6-dimethylphenyl)-trifluoro-$l^{4}-sulfane |
| SMILES | CC1=CC(=CC(=C1S(F)(F)F)C)C(C)(C)C |
Chemical Identifiers
| 947725-04-4 | |
| 250.323 | |
| VRTQPEYVMHATOA-UHFFFAOYSA-N | |
| 44184700 | |
| CC1=CC(=CC(=C1S(F)(F)F)C)C(C)(C)C |
| C12H17F3S | |
| MFCD11858620 | |
| FLUOLEAD(TM) | |
| (4-tert-butyl-2,6-dimethylphenyl)-trifluoro-$l^{4}-sulfane |
Specifications
| 947725-04-4 | |
| 150°C | |
| MFCD11858620 | |
| 3261 | |
| VRTQPEYVMHATOA-UHFFFAOYSA-N | |
| (4-tert-butyl-2,6-dimethylphenyl)-trifluoro-$l^{4}-sulfane | |
| 44184700 | |
| ≥90.0% (T) | |
| 4-tert-Butyl-2,6-dimethylphenylsulfur Trifluoride |
| 63°C | |
| C12H17F3S | |
| 1 g | |
| FLUOLEAD(TM) | |
| CC1=CC(=CC(=C1S(F)(F)F)C)C(C)(C)C | |
| 250.323 | |
| 250.32 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator