missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl-2,6-dimethylphenylsulfur Trifluoride 90.0+%, TCI America™

Catalog No. B36641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36641G 1 g
B36645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36641G Supplier TCI America Supplier No. B36641G

Chemical Identifiers

CAS 947725-04-4
Molecular Formula C12H17F3S
Molecular Weight (g/mol) 250.323
MDL Number MFCD11858620
InChI Key VRTQPEYVMHATOA-UHFFFAOYSA-N
Synonym FLUOLEAD(TM)
PubChem CID 44184700
IUPAC Name (4-tert-butyl-2,6-dimethylphenyl)-trifluoro-$l^{4}-sulfane
SMILES CC1=CC(=CC(=C1S(F)(F)F)C)C(C)(C)C

Specifications

Melting Point 63°C
Boiling Point 150°C
Quantity 1 g
UN Number 3261
Formula Weight 250.32
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butyl-2,6-dimethylphenylsulfur Trifluoride
TSCA No
Recommended Storage Refrigerator