missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butoxyphenol 98.0+%, TCI America™

Catalog No. B23901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23901G 1 g
B23905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23901G Supplier TCI America Supplier No. B23901G

Chemical Identifiers

CAS 2460-87-9
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD02183556
InChI Key CIICLJLSRUHUBY-UHFFFAOYSA-N
Synonym 4-tert-butoxy phenol, 4-tert-butoxyphenol, p-tert-butoxyphenol, 4-t-butoxyphenol, phenol, 4-1,1-dimethylethoxy, unii-4w56a32ou8, 4-2-methylpropan-2-yl oxy phenol, phenol, p-tert-butoxy, 4-tert-butoxy-phenol, acmc-209gdf
PubChem CID 2773621
IUPAC Name 4-[(2-methylpropan-2-yl)oxy]phenol
SMILES CC(C)(C)OC1=CC=C(C=C1)O

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 1 g
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butoxyphenol