accessibility menu, dialog, popup

UserName

CAS RN 2460-87-9

IUPAC Name: 4-(tert-butoxy)phenol
Synonyms: 4-(tert-butoxy)phenol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: CIICLJLSRUHUBY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)