missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Amylphenol 98.0+%, TCI America™

Catalog No. p-7116914
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
A046025G 25 g
A0460500G 500 g
2 options

Catalog No. A046025G

Supplier: TCI America A046025G

Only null left

Chemical Identifiers

CAS 80-46-6
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00002369
InChI Key NRZWYNLTFLDQQX-UHFFFAOYSA-N
Synonym 4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol
PubChem CID 6643
ChEBI CHEBI:35096
IUPAC Name 4-(2-methylbutan-2-yl)phenol
SMILES CCC(C)(C)C1=CC=C(C=C1)O

Specifications

Melting Point 94°C
Color White-Yellow
Boiling Point 262°C
Quantity 25 g
UN Number 2430
Formula Weight 164.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Amylphenol
EINECSNumber (3)-0503
RTECSNumber SM6825000
TSCA Yes