accessibility menu, dialog, popup

UserName

H3CH3CCH3OH

CAS RN 80-46-6

IUPAC Name: 4-(2-methylbutan-2-yl)phenol
Synonyms: tert-amylphenol caswell no. 050 amilfenol Ptap pentaphen amyl phenol 4T epa pesticide chemical code 064101 amilphenol ucar amyl phenol 4T P-tert-amylphenol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: NRZWYNLTFLDQQX-UHFFFAOYSA-N
SMILES: CCC(C)(C)C1=CC=C(O)C=C1
Boiling Point 
  • (1)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (1)
  • (4)
  • (2)
  • (3)
  • (4)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)