missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Amylphenol, 99%

CAS: 80-46-6 | C11H16O | 164.25 g/mol

Supplier:  Thermo Scientific Chemicals 167332500

 View more versions of this product

Catalog No. AC167332500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-tert-Amylphenol
80-46-6
100.0
White to Yellow
111°C
98.5% min. (GC)
C11H16O
MFCD00002369
06, 548
4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol
NRZWYNLTFLDQQX-UHFFFAOYSA-N
4-(2-methylbutan-2-yl)phenol
6643
CHEBI:35096
99%
99%
98.5
>91.0°C
255.0°C
Authentic
Glass bottle
C2H5C(CH3)2C6H4OH
250 g
15, 7255
Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, benzene and chloroform
CCC(C)(C)C1=CC=C(C=C1)O
164.25
2.9 mPa.s (100°C)
164.25
Briquettes or Flakes to Coarse Powder

Chemical Identifiers

80-46-6
164.25
NRZWYNLTFLDQQX-UHFFFAOYSA-N
6643
4-(2-methylbutan-2-yl)phenol
C11H16O
MFCD00002369
4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol
CHEBI:35096
CCC(C)(C)C1=CC=C(C=C1)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several m

GHS Signal Word: Danger

EINECSNumber : 201-280-9

RTECSNumber : SM6825000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only