missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Sulfophthalic Acid (contains 3-Sulfophthalic Acid) (ca. 50% in Water), TCI America™
$35.41 - $74.56
Chemical Identifiers
| CAS | 89-08-7 |
|---|---|
| Molecular Formula | C8H6O7S |
| Molecular Weight (g/mol) | 246.189 |
| MDL Number | MFCD00007494 |
| InChI Key | WNKQDGLSQUASME-UHFFFAOYSA-N |
| Synonym | 1,2-benzenedicarboxylic acid, 4-sulfo, phthalic acid, 4-sulfo, 4-sulphophthalic acid, unii-t1sm5dom66, 4-sulfobenzene-1,2-dicarboxylic acid, t1sm5dom66, 4-sulfophthalic acid aq. soln., 5-sulfophthalic acid, acmc-209uex, 1, 4-sulfo |
| PubChem CID | 6962 |
| IUPAC Name | 4-sulfophthalic acid |
| SMILES | C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
S013625ML
|
TCI America
S013625ML |
25 mL |
Each for $35.41
|
|
|||||
|
S0136500ML
|
TCI America
S0136500ML |
500 mL |
Each for $74.56
|
|
|||||
Chemical Identifiers
| 89-08-7 | |
| 246.189 | |
| WNKQDGLSQUASME-UHFFFAOYSA-N | |
| 6962 | |
| C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)C(=O)O |
| C8H6O7S | |
| MFCD00007494 | |
| 1,2-benzenedicarboxylic acid, 4-sulfo, phthalic acid, 4-sulfo, 4-sulphophthalic acid, unii-t1sm5dom66, 4-sulfobenzene-1,2-dicarboxylic acid, t1sm5dom66, 4-sulfophthalic acid aq. soln., 5-sulfophthalic acid, acmc-209uex, 1, 4-sulfo | |
| 4-sulfophthalic acid |
Specifications
| 89-08-7 | |
| MFCD00007494 | |
| 3265 | |
| WNKQDGLSQUASME-UHFFFAOYSA-N | |
| 4-sulfophthalic acid | |
| 6962 | |
| Liquid |
| C8H6O7S | |
| 25 mL | |
| 1,2-benzenedicarboxylic acid, 4-sulfo, phthalic acid, 4-sulfo, 4-sulphophthalic acid, unii-t1sm5dom66, 4-sulfobenzene-1,2-dicarboxylic acid, t1sm5dom66, 4-sulfophthalic acid aq. soln., 5-sulfophthalic acid, acmc-209uex, 1, 4-sulfo | |
| C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)C(=O)O | |
| 246.189 | |
| 246.19 | |
| 4-Sulfophthalic Acid (contains 3-Sulfophthalic Acid) (ca. 50% in Water) |
Safety and Handling
EINECSNumber : (3)-2131
TSCA : Yes