accessibility menu, dialog, popup

UserName

CAS RN 89-08-7

IUPAC Name: 4-sulfobenzene-1,2-dicarboxylic acid
Synonyms: 4-sulfobenzene-1,2-dicarboxylic acid
Molecular Weight (g/mol): 246.19
Molecular Formula: C8H6O7S
InChi Key: WNKQDGLSQUASME-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1C(O)=O)S(O)(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results