missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfophthalic Acid (contains 3-Sulfophthalic Acid) (ca. 50% in Water), TCI America™

Catalog No. S013625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S013625ML 25 mL
S0136500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S013625ML Supplier TCI America Supplier No. S013625ML

Chemical Identifiers

CAS 89-08-7
Molecular Formula C8H6O7S
Molecular Weight (g/mol) 246.189
MDL Number MFCD00007494
InChI Key WNKQDGLSQUASME-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, 4-sulfo, phthalic acid, 4-sulfo, 4-sulphophthalic acid, unii-t1sm5dom66, 4-sulfobenzene-1,2-dicarboxylic acid, t1sm5dom66, 4-sulfophthalic acid aq. soln., 5-sulfophthalic acid, acmc-209uex, 1, 4-sulfo
PubChem CID 6962
IUPAC Name 4-sulfophthalic acid
SMILES C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)C(=O)O

Specifications

Quantity 25 mL
UN Number 3265
Formula Weight 246.19
Physical Form Liquid
Chemical Name or Material 4-Sulfophthalic Acid (contains 3-Sulfophthalic Acid) (ca. 50% in Water)
EINECSNumber (3)-2131
TSCA Yes