Learn More
4-Sulfonamidophenylhydrazine hydrochloride, 97%
CAS: 17852-52-7 | C6H10ClN3O2S | 223.675 g/mol
Supplier: Thermo Scientific Chemicals B2001518
Description
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 4-Sulfonamidophenylhydrazine hydrochloride | |
| 217°C to 219°C (decomposition) | |
| MFCD00052262 | |
| Hygroscopic | |
| Soluble in water. | |
| C1=CC(=CC=C1NN)S(=O)(=O)N.Cl | |
| 223.675 | |
| 223.68 |
| 17852-52-7 | |
| C6H10ClN3O2S | |
| 50 g | |
| 4-hydrazinylbenzenesulfonamide hydrochloride, 4-hydrazinobenzene-1-sulfonamide hydrochloride, 4-hydrazinobenzenesulfonamide hydrochloride, 4-sulfonamidephenylhydrazine hcl, 4-aminosulfonylphenylhydrazine hydrochloride, 4-hydrazinylbenzenesulfonamide hydrochloride 1:x, 4-sulfamoylphenyl hydrazine hydrochloride, 4-sulfonamide-phenylhydrazine hcl, phenylhydrazine-4-sulfonamide hydrochloride | |
| IKEURONJLPUALY-UHFFFAOYSA-N | |
| 4-hydrazinylbenzenesulfonamide;hydrochloride | |
| 2794567 | |
| 97% |
Chemical Identifiers
| 17852-52-7 | |
| 223.675 | |
| IKEURONJLPUALY-UHFFFAOYSA-N | |
| 2794567 | |
| C1=CC(=CC=C1NN)S(=O)(=O)N.Cl |
| C6H10ClN3O2S | |
| MFCD00052262 | |
| 4-hydrazinylbenzenesulfonamide hydrochloride, 4-hydrazinobenzene-1-sulfonamide hydrochloride, 4-hydrazinobenzenesulfonamide hydrochloride, 4-sulfonamidephenylhydrazine hcl, 4-aminosulfonylphenylhydrazine hydrochloride, 4-hydrazinylbenzenesulfonamide hydrochloride 1:x, 4-sulfamoylphenyl hydrazine hydrochloride, 4-sulfonamide-phenylhydrazine hcl, phenylhydrazine-4-sulfonamide hydrochloride | |
| 4-hydrazinylbenzenesulfonamide;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only