missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Sulfocalix[8]arene Hydrate 98.0+%, TCI America™
Supplier: TCI America S04715G
Specifications
| 4-Sulfocalix[8]arene Hydrate | |
| C56H48O32S8 | |
| 5 g | |
| OC1=C2CC3=CC(=CC(CC4=CC(=CC(CC5=CC(=CC(CC6=CC(=CC(CC7=CC(=CC(CC8=CC(=CC(CC9=CC(=CC(CC1=CC(=C2)S(O)(=O)=O)=C9O)S(O)(=O)=O)=C8O)S(O)(=O)=O)=C7O)S(O)(=O)=O)=C6O)S(O)(=O)=O)=C5O)S(O)(=O)=O)=C4O)S(O)(=O)=O)=C3O)S(O)(=O)=O | |
| 1489.45 | |
| 1489.45 | |
| Crystalline Powder |
| 137407-62-6 | |
| MFCD00917032 | |
| KCEGJGDGMRAJEP-UHFFFAOYSA-N | |
| 49,50,51,52,53,54,55,56-octahydroxynonacyclo[43.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹.1²¹,²âµ.1²â·,³¹.1³³,³â·.1³â¹,â´Â³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-5,11,17,23,29,35,41,47-octasulfonic acid | |
| 644085 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 137407-62-6 | |
| 1489.45 | |
| KCEGJGDGMRAJEP-UHFFFAOYSA-N | |
| 49,50,51,52,53,54,55,56-octahydroxynonacyclo[43.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹.1²¹,²âµ.1²â·,³¹.1³³,³â·.1³â¹,â´Â³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-5,11,17,23,29,35,41,47-octasulfonic acid |
| C56H48O32S8 | |
| MFCD00917032 | |
| 644085 | |
| OC1=C2CC3=CC(=CC(CC4=CC(=CC(CC5=CC(=CC(CC6=CC(=CC(CC7=CC(=CC(CC8=CC(=CC(CC9=CC(=CC(CC1=CC(=C2)S(O)(=O)=O)=C9O)S(O)(=O)=O)=C8O)S(O)(=O)=O)=C7O)S(O)(=O)=O)=C6O)S(O)(=O)=O)=C5O)S(O)(=O)=O)=C4O)S(O)(=O)=O)=C3O)S(O)(=O)=O |
Safety and Handling
TSCA : No