missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfocalix[8]arene Hydrate 98.0+%, TCI America™

Catalog No. p-7135899
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04711G 1 g
S04715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04711G Supplier TCI America Supplier No. S04711G
Only null left

Chemical Identifiers

CAS 137407-62-6
Molecular Formula C56H48O32S8
Molecular Weight (g/mol) 1489.45
MDL Number MFCD00917032
InChI Key KCEGJGDGMRAJEP-UHFFFAOYSA-N
PubChem CID 644085
IUPAC Name 49,50,51,52,53,54,55,56-octahydroxynonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-5,11,17,23,29,35,41,47-octasulfonic acid
SMILES OC1=C2CC3=CC(=CC(CC4=CC(=CC(CC5=CC(=CC(CC6=CC(=CC(CC7=CC(=CC(CC8=CC(=CC(CC9=CC(=CC(CC1=CC(=C2)S(O)(=O)=O)=C9O)S(O)(=O)=O)=C8O)S(O)(=O)=O)=C7O)S(O)(=O)=O)=C6O)S(O)(=O)=O)=C5O)S(O)(=O)=O)=C4O)S(O)(=O)=O)=C3O)S(O)(=O)=O

Specifications

Quantity 1 g
Formula Weight 1489.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Sulfocalix[8]arene Hydrate