CAS RN 137407-62-6
CAS RN 137407-62-6
IUPAC Name:
49,50,51,52,53,54,55,56-octahydroxynonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-5,11,17,23,29,35,41,47-octasulfonic acid
Synonyms:
49,50,51,52,53,54,55,56-octahydroxynonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-5,11,17,23,29,35,41,47-octasulfonic acid
Molecular Weight (g/mol):
1489.45
Molecular Formula:
C56H48O32S8
InChi Key:
KCEGJGDGMRAJEP-UHFFFAOYSA-N
SMILES:
OC1=C2CC3=CC(=CC(CC4=CC(=CC(CC5=CC(=CC(CC6=CC(=CC(CC7=CC(=CC(CC8=CC(=CC(CC9=CC(=CC(CC1=CC(=C2)S(O)(=O)=O)=C9O)S(O)(=O)=O)=C8O)S(O)(=O)=O)=C7O)S(O)(=O)=O)=C6O)S(O)(=O)=O)=C5O)S(O)(=O)=O)=C4O)S(O)(=O)=O)=C3O)S(O)(=O)=O
1
–
1
of
1
results
1
4-Sulfocalix[8]arene Hydrate 98.0+%, TCI America™
CAS: 137407-62-6 Molecular Formula: C56H48O32S8 Molecular Weight (g/mol): 1489.45 MDL Number: MFCD00917032 InChI Key: KCEGJGDGMRAJEP-UHFFFAOYSA-N PubChem CID: 644085 IUPAC Name: 49,50,51,52,53,54,55,56-octahydroxynonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-5,11,17,23,29,35,41,47-octasulfonic acid SMILES: OC1=C2CC3=CC(=CC(CC4=CC(=CC(CC5=CC(=CC(CC6=CC(=CC(CC7=CC(=CC(CC8=CC(=CC(CC9=CC(=CC(CC1=CC(=C2)S(O)(=O)=O)=C9O)S(O)(=O)=O)=C8O)S(O)(=O)=O)=C7O)S(O)(=O)=O)=C6O)S(O)(=O)=O)=C5O)S(O)(=O)=O)=C4O)S(O)(=O)=O)=C3O)S(O)(=O)=O