missing translation for 'onlineSavingsMsg'
Learn More

4-sec-Butyl-2,6-di-tert-butylphenol 98.0+%, TCI America™

Catalog No. B277425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B277425G 25 g
B2774500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B277425G Supplier TCI America Supplier No. B277425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17540-75-9
Molecular Formula C18H30O
Molecular Weight (g/mol) 262.437
InChI Key BFZOTKYPSZSDEV-UHFFFAOYSA-N
PubChem CID 86583
IUPAC Name 4-butan-2-yl-2,6-ditert-butylphenol
SMILES CCC(C)C1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

Color White-Yellow
Boiling Point 141°C
Quantity 25 g
Formula Weight 262.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-sec-Butyl-2,6-di-tert-butylphenol
EINECSNumber (3)-0540
TSCA Yes