accessibility menu, dialog, popup

UserName

CH3CH3OHH3CH3CCH3CH3H3CCH3

CAS RN 17540-75-9

IUPAC Name: 4-(butan-2-yl)-2,6-di-tert-butylphenol
Synonyms: 2,6-di-tert-butyl-4-(sec-butyl)phenol
Molecular Weight (g/mol): 262.44
Molecular Formula: C18H30O
InChi Key: BFZOTKYPSZSDEV-UHFFFAOYNA-N
SMILES: CCC(C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results