missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Quinolinecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America Q006525G
Specifications
| 4-Quinolinecarboxylic Acid | |
| White-Yellow | |
| MFCD00006782 | |
| 4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure | |
| C1=CC=C2C(=C1)C(=CC=N2)C(=O)O | |
| 173.171 | |
| CHEBI:18311 | |
| ≥98.0% (HPLC,T) |
| 486-74-8 | |
| C10H7NO2 | |
| 25 g | |
| VQMSRUREDGBWKT-UHFFFAOYSA-N | |
| quinoline-4-carboxylic acid | |
| 10243 | |
| 173.17 | |
| Crystalline Powder |
Chemical Identifiers
| 486-74-8 | |
| 173.171 | |
| VQMSRUREDGBWKT-UHFFFAOYSA-N | |
| 10243 | |
| quinoline-4-carboxylic acid |
| C10H7NO2 | |
| MFCD00006782 | |
| 4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure | |
| CHEBI:18311 | |
| C1=CC=C2C(=C1)C(=CC=N2)C(=O)O |
Safety and Handling
RTECSNumber : GD3850000
TSCA : No