accessibility menu, dialog, popup

UserName

HOON

CAS RN 486-74-8

IUPAC Name: quinoline-4-carboxylic acid
Synonyms: quinoline-4-carboxylic acid usaf D-2
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: VQMSRUREDGBWKT-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=NC=C1
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)