missing translation for 'onlineSavingsMsg'
Learn More

4-Quinolinecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7135636
Change view
Click to view available options
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
Q00651G 1 g
Q00655G 5 g
Q006525G 25 g
3 options

Catalog No. Q00651G

Supplier: TCI America Q00651G

Only null left

Chemical Identifiers

CAS 486-74-8
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00006782
InChI Key VQMSRUREDGBWKT-UHFFFAOYSA-N
Synonym 4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure
PubChem CID 10243
ChEBI CHEBI:18311
IUPAC Name quinoline-4-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC=N2)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 173.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Quinolinecarboxylic Acid
RTECSNumber GD3850000
TSCA No