missing translation for 'onlineSavingsMsg'
Learn More

4-(Pyrrolidin-1-ylmethyl)benzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 159691-25-5
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
InChI Key DOEIPZUTRVMWAB-UHFFFAOYSA-N
Synonym 4-pyrrolidin-1-ylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoic acid, 4-pyrrolidin-1-yl methyl benzoic acid, 4-pyrrolidinylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoicacid, 4-1-pyrrolidinylmethyl-benzoic acid, benzoic acid, 4-1-pyrrolidinylmethyl-, hydrochloride
PubChem CID 1092976
IUPAC Name 4-(pyrrolidin-1-ylmethyl)benzoic acid
SMILES C1CCN(C1)CC2=CC=C(C=C2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC55601CB
View Documents Promotion Details
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC55601DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC55601EA
View Documents Promotion Details
N/A 10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

159691-25-5
205.257
4-pyrrolidin-1-ylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoic acid, 4-pyrrolidin-1-yl methyl benzoic acid, 4-pyrrolidinylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoicacid, 4-1-pyrrolidinylmethyl-benzoic acid, benzoic acid, 4-1-pyrrolidinylmethyl-, hydrochloride
4-(pyrrolidin-1-ylmethyl)benzoic acid
C12H15NO2
DOEIPZUTRVMWAB-UHFFFAOYSA-N
1092976
C1CCN(C1)CC2=CC=C(C=C2)C(=O)O

Specifications

159691-25-5
C12H15NO2
DOEIPZUTRVMWAB-UHFFFAOYSA-N
4-(pyrrolidin-1-ylmethyl)benzoic acid
1092976
97%
97%
4-pyrrolidin-1-ylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoic acid, 4-pyrrolidin-1-yl methyl benzoic acid, 4-pyrrolidinylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoicacid, 4-1-pyrrolidinylmethyl-benzoic acid, benzoic acid, 4-1-pyrrolidinylmethyl-, hydrochloride
C1CCN(C1)CC2=CC=C(C=C2)C(=O)O
205.257
205.26
4-(Pyrrolidin-1-ylmethyl)benzoic acid
Videos
SDS
Documents

Documents