missing translation for 'onlineSavingsMsg'
Learn More

4-(Pyrrolidin-1-ylmethyl)benzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC55601EA

Quantity 10g

 View more versions of this product

Catalog No. CC55601EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

159691-25-5
205.257
4-pyrrolidin-1-ylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoic acid, 4-pyrrolidin-1-yl methyl benzoic acid, 4-pyrrolidinylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoicacid, 4-1-pyrrolidinylmethyl-benzoic acid, benzoic acid, 4-1-pyrrolidinylmethyl-, hydrochloride
4-(pyrrolidin-1-ylmethyl)benzoic acid
C12H15NO2
DOEIPZUTRVMWAB-UHFFFAOYSA-N
1092976
C1CCN(C1)CC2=CC=C(C=C2)C(=O)O

Specifications

4-(Pyrrolidin-1-ylmethyl)benzoic acid
97%
C12H15NO2
4-pyrrolidin-1-ylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoic acid, 4-pyrrolidin-1-yl methyl benzoic acid, 4-pyrrolidinylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoicacid, 4-1-pyrrolidinylmethyl-benzoic acid, benzoic acid, 4-1-pyrrolidinylmethyl-, hydrochloride
C1CCN(C1)CC2=CC=C(C=C2)C(=O)O
205.257
205.26
159691-25-5
Amber Glass Bottle
10g
DOEIPZUTRVMWAB-UHFFFAOYSA-N
4-(pyrrolidin-1-ylmethyl)benzoic acid
1092976
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents