missing translation for 'onlineSavingsMsg'
Learn More

(4-Pyridylthio)acetic Acid 98.0+%, TCI America™

Catalog No. P100025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P100025G 25 g
P1000500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P100025G Supplier TCI America Supplier No. P100025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10351-19-6
Molecular Formula C7H7NO2S
Molecular Weight (g/mol) 169.198
MDL Number MFCD00006424
InChI Key PGUPJAPHYIEKLT-UHFFFAOYSA-N
Synonym 4-pyridylthio acetic acid, 2-pyridin-4-ylthio acetic acid, acetic acid, 4-pyridinylthio, 4-pyridylthioacetic acid, pyridin-4-ylsulfanyl acetic acid, 2-pyridin-4-ylsulfanyl acetic acid, 4-pyridinylthio acetic acid, acetic acid, 2-4-pyridinylthio, s-4-pyridyl thioglycolic acid, 2-4-pyridylthio acetic acid
PubChem CID 82574
IUPAC Name 2-pyridin-4-ylsulfanylacetic acid
SMILES C1=CN=CC=C1SCC(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 169.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Pyridylthio)acetic Acid