CAS RN 10351-19-6
IUPAC Name:
2-(pyridin-4-ylsulfanyl)acetate
Synonyms:
2-(pyridin-4-ylsulfanyl)acetate
Molecular Weight (g/mol):
168.19
Molecular Formula:
C7H6NO2S
InChi Key:
PGUPJAPHYIEKLT-UHFFFAOYSA-M
SMILES:
[O-]C(=O)CSC1=CC=NC=C1
1
–
1
of
1
results
1
(4-Pyridylthio)acetic Acid 98.0+%, TCI America™
CAS: 10351-19-6 Molecular Formula: C7H7NO2S Molecular Weight (g/mol): 169.198 MDL Number: MFCD00006424 InChI Key: PGUPJAPHYIEKLT-UHFFFAOYSA-N Synonym: 4-pyridylthio acetic acid,2-pyridin-4-ylthio acetic acid,acetic acid, 4-pyridinylthio,4-pyridylthioacetic acid,pyridin-4-ylsulfanyl acetic acid,2-pyridin-4-ylsulfanyl acetic acid,4-pyridinylthio acetic acid,acetic acid, 2-4-pyridinylthio,s-4-pyridyl thioglycolic acid,2-4-pyridylthio acetic acid PubChem CID: 82574 IUPAC Name: 2-pyridin-4-ylsulfanylacetic acid SMILES: C1=CN=CC=C1SCC(=O)O