missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pyridylacetic Acid Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P131725G
Specifications
| 4-Pyridylacetic Acid Hydrochloride | |
| White-Yellow | |
| MFCD00012827 | |
| 4-pyridineacetic acid hydrochloride, 4-pyridylacetic acid hydrochloride, 2-pyridin-4-yl acetic acid hydrochloride, 4-carboxymethylpyridinium chloride, 4-acetic acid pyridine hydrochloride, 4-pyridineacetic acid hcl, 4-pyridine acetic acid hydrochloride, 2-4-pyridyl acetic acid hydrochloride, 4-acetylpyridine hydrochloride, 4-pyridineacetic acid, hydrochloride | |
| C1=CN=CC=C1CC(=O)O.Cl | |
| 173.596 | |
| 173.60 | |
| Crystalline Powder |
| 6622-91-9 | |
| C7H8ClNO2 | |
| 25 g | |
| WKJRYVOTVRPAFN-UHFFFAOYSA-N | |
| 2-pyridin-4-ylacetic acid;hydrochloride | |
| 81097 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 6622-91-9 | |
| 173.596 | |
| WKJRYVOTVRPAFN-UHFFFAOYSA-N | |
| 81097 | |
| C1=CN=CC=C1CC(=O)O.Cl |
| C7H8ClNO2 | |
| MFCD00012827 | |
| 4-pyridineacetic acid hydrochloride, 4-pyridylacetic acid hydrochloride, 2-pyridin-4-yl acetic acid hydrochloride, 4-carboxymethylpyridinium chloride, 4-acetic acid pyridine hydrochloride, 4-pyridineacetic acid hcl, 4-pyridine acetic acid hydrochloride, 2-4-pyridyl acetic acid hydrochloride, 4-acetylpyridine hydrochloride, 4-pyridineacetic acid, hydrochloride | |
| 2-pyridin-4-ylacetic acid;hydrochloride |
Safety and Handling
TSCA : No