missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridylacetic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. p-7135073
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13175G 5 g
P131725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13175G Supplier TCI America Supplier No. P13175G

Chemical Identifiers

CAS 6622-91-9
Molecular Formula C7H8ClNO2
Molecular Weight (g/mol) 173.596
MDL Number MFCD00012827
InChI Key WKJRYVOTVRPAFN-UHFFFAOYSA-N
Synonym 4-pyridineacetic acid hydrochloride, 4-pyridylacetic acid hydrochloride, 2-pyridin-4-yl acetic acid hydrochloride, 4-carboxymethylpyridinium chloride, 4-acetic acid pyridine hydrochloride, 4-pyridineacetic acid hcl, 4-pyridine acetic acid hydrochloride, 2-4-pyridyl acetic acid hydrochloride, 4-acetylpyridine hydrochloride, 4-pyridineacetic acid, hydrochloride
PubChem CID 81097
IUPAC Name 2-pyridin-4-ylacetic acid;hydrochloride
SMILES C1=CN=CC=C1CC(=O)O.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 173.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Pyridylacetic Acid Hydrochloride