accessibility menu, dialog, popup

UserName

CAS RN 6622-91-9

IUPAC Name: 2-(pyridin-4-yl)acetic acid hydrochloridyl
Synonyms: 4-acetylpyridine hydrochloridyl
Molecular Weight (g/mol): 172.59
Molecular Formula: C7H7ClNO2
InChi Key: NDCASQYPYLKYOP-UHFFFAOYSA-N
SMILES: [Cl].OC(=O)CC1=CC=NC=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)