Learn More
4-Pyridineacetic acid hydrochloride, 98+%
CAS: 6622-91-9 | C7H8ClNO2 | 173.596 g/mol
Supplier: Thermo Scientific Chemicals A1434814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Pyridineacetic acid hydrochloride | |
| ∼136°C (decomposition) | |
| C7H8ClNO2 | |
| 25 g | |
| 4-pyridineacetic acid hydrochloride, 4-pyridylacetic acid hydrochloride, 2-pyridin-4-yl acetic acid hydrochloride, 4-carboxymethylpyridinium chloride, 4-acetic acid pyridine hydrochloride, 4-pyridineacetic acid hcl, 4-pyridine acetic acid hydrochloride, 2-4-pyridyl acetic acid hydrochloride, 4-acetylpyridine hydrochloride, 4-pyridineacetic acid, hydrochloride | |
| C1=CN=CC=C1CC(=O)O.Cl | |
| 173.596 | |
| 173.6 |
| 6622-91-9 | |
| Odorless | |
| MFCD00012827 | |
| 3696648 | |
| WKJRYVOTVRPAFN-UHFFFAOYSA-N | |
| 2-pyridin-4-ylacetic acid;hydrochloride | |
| 81097 | |
| ≥98% |
Chemical Identifiers
| 6622-91-9 | |
| 173.596 | |
| WKJRYVOTVRPAFN-UHFFFAOYSA-N | |
| 81097 | |
| C1=CN=CC=C1CC(=O)O.Cl |
| C7H8ClNO2 | |
| MFCD00012827 | |
| 4-pyridineacetic acid hydrochloride, 4-pyridylacetic acid hydrochloride, 2-pyridin-4-yl acetic acid hydrochloride, 4-carboxymethylpyridinium chloride, 4-acetic acid pyridine hydrochloride, 4-pyridineacetic acid hcl, 4-pyridine acetic acid hydrochloride, 2-4-pyridyl acetic acid hydrochloride, 4-acetylpyridine hydrochloride, 4-pyridineacetic acid, hydrochloride | |
| 2-pyridin-4-ylacetic acid;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 229-576-3
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only