missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridineacetic acid hydrochloride, 98+%

Catalog No. p-7021842
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1434806 5 g
AAA1434814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1434806 Supplier Thermo Scientific Chemicals Supplier No. A1434806
Only null left

CAS: 6622-91-9 | C7H8ClNO2 | 173.596 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6622-91-9
Molecular Formula C7H8ClNO2
Molecular Weight (g/mol) 173.596
MDL Number MFCD00012827
InChI Key WKJRYVOTVRPAFN-UHFFFAOYSA-N
Synonym 4-pyridineacetic acid hydrochloride, 4-pyridylacetic acid hydrochloride, 2-pyridin-4-yl acetic acid hydrochloride, 4-carboxymethylpyridinium chloride, 4-acetic acid pyridine hydrochloride, 4-pyridineacetic acid hcl, 4-pyridine acetic acid hydrochloride, 2-4-pyridyl acetic acid hydrochloride, 4-acetylpyridine hydrochloride, 4-pyridineacetic acid, hydrochloride
PubChem CID 81097
IUPAC Name 2-pyridin-4-ylacetic acid;hydrochloride
SMILES C1=CN=CC=C1CC(=O)O.Cl

Specifications

Melting Point ∼136°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3696648
Formula Weight 173.6
Percent Purity ≥98%
Chemical Name or Material 4-Pyridineacetic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-576-3
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only