missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Propyl-4-biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$68.97 - $68.97
Specifications
| CAS | 153035-56-4 |
|---|---|
| Color | White-Yellow |
| Molecular Formula | C15H17BO2 |
| MDL Number | MFCD01114670 |
| InChI Key | NOQFUISBLHCDSR-UHFFFAOYSA-N |
Specifications
| 153035-56-4 | |
| C15H17BO2 | |
| NOQFUISBLHCDSR-UHFFFAOYSA-N | |
| [4-(4-propylphenyl)phenyl]boronic acid | |
| 19042596 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD01114670 | |
| B(C1=CC=C(C=C1)C2=CC=C(C=C2)CCC)(O)O | |
| 240.109 | |
| 240.11 |