accessibility menu, dialog, popup

UserName

CAS RN 153035-56-4

BCH3HOOH

CAS RN 153035-56-4

IUPAC Name: {4'-propyl-[1,1'-biphenyl]-4-yl}boronic acid
Synonyms: 4'-propyl-[1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol): 240.11
Molecular Formula: C15H17BO2
InChi Key: NOQFUISBLHCDSR-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)C1=CC=C(C=C1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results