missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylpyridine N-Oxide 98.0+%, TCI America™
Supplier: TCI America P140625G
Specifications
| 4-Phenylpyridine N-Oxide | |
| 155°C | |
| MFCD00006208 | |
| VZOPVKZLLGMDDG-UHFFFAOYSA-N | |
| 1-oxido-4-phenylpyridin-1-ium | |
| 70803 | |
| ≥98.0% (HPLC,T) |
| 1131-61-9 | |
| C11H9NO | |
| 25 g | |
| C1=CC=C(C=C1)C2=CC=[N+](C=C2)[O-] | |
| 171.199 | |
| 171.20 | |
| Crystalline Powder |
Chemical Identifiers
| 1131-61-9 | |
| 171.199 | |
| VZOPVKZLLGMDDG-UHFFFAOYSA-N | |
| 1-oxido-4-phenylpyridin-1-ium |
| C11H9NO | |
| MFCD00006208 | |
| 70803 | |
| C1=CC=C(C=C1)C2=CC=[N+](C=C2)[O-] |
Safety and Handling
TSCA : Yes