CAS RN 1131-61-9
CAS RN 1131-61-9
IUPAC Name:
4-phenylpyridin-1-ium-1-olate
Synonyms:
4-phenylpyridine N-oxide
Molecular Weight (g/mol):
171.2
Molecular Formula:
C11H9NO
InChi Key:
VZOPVKZLLGMDDG-UHFFFAOYSA-N
SMILES:
[O-][N+]1=CC=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Phenylpyridine N-Oxide 98.0+%, TCI America™
CAS: 1131-61-9 Molecular Formula: C11H9NO Molecular Weight (g/mol): 171.199 MDL Number: MFCD00006208 InChI Key: VZOPVKZLLGMDDG-UHFFFAOYSA-N PubChem CID: 70803 IUPAC Name: 1-oxido-4-phenylpyridin-1-ium SMILES: C1=CC=C(C=C1)C2=CC=[N+](C=C2)[O-]