missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylpyridine N-Oxide 98.0+%, TCI America™

Catalog No. p-7135121
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14065G 5 g
P140625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14065G Supplier TCI America Supplier No. P14065G

Chemical Identifiers

CAS 1131-61-9
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.199
MDL Number MFCD00006208
InChI Key VZOPVKZLLGMDDG-UHFFFAOYSA-N
PubChem CID 70803
IUPAC Name 1-oxido-4-phenylpyridin-1-ium
SMILES C1=CC=C(C=C1)C2=CC=[N+](C=C2)[O-]

Specifications

Melting Point 155°C
Quantity 5 g
Formula Weight 171.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylpyridine N-Oxide