missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylbutyric Acid 98.0+%, TCI America™
Supplier: TCI America P064325G
Specifications
| 4-Phenylbutyric Acid | |
| 52°C | |
| 175°C | |
| MFCD00004403 | |
| 4-phenylbutyric acid, benzenebutanoic acid, benzenebutyric acid, phenylbutyrate, gamma-phenylbutyric acid, omega-phenylbutanoic acid, phenylbutyric acid, 4-phenylbutyrate, butyric acid, 4-phenyl, 4-phenyl-n-butyric acid | |
| C1=CC=C(C=C1)CCCC(=O)O | |
| 164.204 | |
| CHEBI:41500 | |
| ≥98.0% (GC,T) |
| 1821-12-1 | |
| White-Yellow | |
| C10H12O2 | |
| 25 g | |
| OBKXEAXTFZPCHS-UHFFFAOYSA-N | |
| 4-phenylbutanoic acid | |
| 4775 | |
| 164.20 | |
| Crystalline Powder |
Chemical Identifiers
| 1821-12-1 | |
| 164.204 | |
| OBKXEAXTFZPCHS-UHFFFAOYSA-N | |
| 4775 | |
| 4-phenylbutanoic acid |
| C10H12O2 | |
| MFCD00004403 | |
| 4-phenylbutyric acid, benzenebutanoic acid, benzenebutyric acid, phenylbutyrate, gamma-phenylbutyric acid, omega-phenylbutanoic acid, phenylbutyric acid, 4-phenylbutyrate, butyric acid, 4-phenyl, 4-phenyl-n-butyric acid | |
| CHEBI:41500 | |
| C1=CC=C(C=C1)CCCC(=O)O |
Safety and Handling
TSCA : Yes