missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyric Acid 98.0+%, TCI America™

Catalog No. p-7134711
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
P064325G 25 g
P0643250G 250 g
2 options

Catalog No. P064325G

Supplier: TCI America P064325G

Only null left

Chemical Identifiers

CAS 1821-12-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00004403
InChI Key OBKXEAXTFZPCHS-UHFFFAOYSA-N
Synonym 4-phenylbutyric acid, benzenebutanoic acid, benzenebutyric acid, phenylbutyrate, gamma-phenylbutyric acid, omega-phenylbutanoic acid, phenylbutyric acid, 4-phenylbutyrate, butyric acid, 4-phenyl, 4-phenyl-n-butyric acid
PubChem CID 4775
ChEBI CHEBI:41500
IUPAC Name 4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCCC(=O)O

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 175°C
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylbutyric Acid