accessibility menu, dialog, popup

UserName

CAS RN 1821-12-1

IUPAC Name: 4-phenylbutanoic acid
Synonyms: 4-phenylbutyric acid 1-phenylbutyric acid g-phenylbutyric acid w-phenylbutanoate buphenyl PBA ammonaps ω-phenylbutanoic acid phenylbutyrate sodium preparation g-phenylbutyrate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: OBKXEAXTFZPCHS-UHFFFAOYSA-N
SMILES: OC(=O)CCCC1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (5)
  • (1)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)